Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Beta-Hydroxybutyrate LiquiColor™ Test (Endpoint)
GSA_VASDP

Catalog No. SBHR100 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
1 x 50mL R1, 1 x 8.5mL R2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SBHR100 1 x 50mL R1, 1 x 8.5mL R2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SBHR100 Supplier Stanbio™ Supplier No. 2440058
Add to Cart
Edge
Add to Cart

Designed for maximum stability and ease-of-use

  • Enzymatic beta-hydroxybutyrate reaction
  • Ready-to-use liquid reagents
  • Use serum or plasma
  • Linear up to 4.5 mmol/L
  • Read at 505 nm

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CE Marked
Type beta-Hydroxybutyrate Test Kit
For Use With (Application) Quantitative testing
Includes 1 x 50mL R1, 1 x 8.5mL R2, 1 x 3mL (1mM/L) Standard
Quantity 1 x 50mL R1, 1 x 8.5mL R2
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.