Learn More
Stanbio™ Quicktest RPR
Shop All Stanbio Laboratory Products
Click to view available options
Quantity:
100 Tests
3 x 5 mL
500 Tests
Type:
QuickTest RPR Test
QuickTest RPR Test Control
Description
Includes
RPR antigen suspension, cardiolipin suspension, RPR reactive control, RPR weakly-reactive control, RPR non-reactive control
- For qualitative and quantitative determination of reagin antibodies in serum or plasma
- Macroscopic non-treponemal flocculation test that is used to detect and quantify reagin, an anti-lipoidal antibody found in serum or plasma from persons with syphilis and occasionally in persons with acute or chronic infections or conditions other than syphilis
- Antigen is modification of VDRL antigen which contains microparticulate charcoal to enhance the visual difference between a reactive and non-reactive result
Specifications
Specifications
Detectable Analytes | Reagin (Syphilis) |
Type | QuickTest RPR Test |
Includes | RPR Antigen suspension, cardiolipin suspension, RPR reactive control, RPR weakly-reactive control, RPR non-reactive control |
Quantity | 500 Tests |