Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ QuPID™ hCG Test
SDP

Catalog No. p-2855971
Click to view available options
Quantity:
25 Tests
50 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy

Qualitative platform immunoassay device

  • Dip and read strip format for the visual qualitative detection of hCG in urine
  • Employs a test strip with a unique combination of monoclonal/polyclonal anti-hCG antibody, a colored reagent and two immobilized capture antibodies directed against hCG and mouse IgG respectively
  • Results in 3 minutes
  • Sensitive to 20 mIU/mL hCG in urine
  • Store at room temperature (2–30°C)

Specifications

Accuracy >99%
Clia Complexity Waived
Detectable Analytes hCG
Sample Type Urine
Type hCG Pregnancy Test Kit
Quantity 25 Tests
Test Time 3 min.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.