Learn More
Stanbio™ QuPID™ Plus hCG Test
SDP
Catalog No. p-2855975
Click to view available options
Quantity:
25 Tests
50 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
Disposable plastic dropper
Qualitative platform immunoassay device
  • Detects hCG in 3 minutes in urine or 5 minutes in serum
  • Sensitive to 20 mIU/mL or greater
  • Specific monoclonal/polyclonal antibodies ensure early pregnancy detection while preventing cross reactivity with hormones present in patient samples
  • Complete instructions printed directly on each individual test unit's foil pouch
Specifications
Accuracy >99%
Clia Complexity Waived
Detectable Analytes hCG
Sample Type Serum, Urine
Type hCG Pregnancy Test
Includes Complete instructions printed directly on each individual test unit's foil pouch, as well as a disposable Plastic dropper for convenience and ease-of-use
Quantity 25 Tests
Test Time 3 min. for urine, 5 min. for serum
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.