Learn More
Caframo™ System Stand for Overhead Stirrers
System Stands allow support for additional apparatus. Wide base and H configuration gives superior stability when agitating viscous materials. For use with Caframo High Torque and Light Torque Overhead Stirrers
Manufacturer:  Caframo™ A114
Catalog No. 14-501-089
Add to cart
Description
Description
  • Wide H base offers excellent stability.
  • Available in three heights to accommodate varying vessel sizes (14-501-087, 14-501-088, 14-501-089).
  • Base composed of cast zinc-aluminum alloy coated with chemical resistant epoxy paint.
  • Threaded holes in base for installation of A300 rods to accommodate additional apparatus.
Specifications
Specifications
Chemical-Resistant Cast Zinc- Aluminum Alloy
Cast zinc-aluminum alloy coated with chemical resistant epoxy pain
25 mm
48 in.
H Base and Rod
416 x 430 x 1219 mm
1 in.
Caframo™ and Fisherbrand™ Overhead Stirrers
1219 mm