Learn More
Thermo Scientific™ Standard Fluid Transfer Assembly Extension Set
Simplify process connectivity and enable greater efficiency for most upstream and downstream process operations.
$24.75 - $138.00
Specifications
Content And Storage | Approved for storage at ambient conditions (15°C to 30°C). |
---|
Catalog Number | Mfr. No. | Tubing Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Tubing Style | Price | Quantity & Availability | |||||
SH3108303
![]() |
Thermo Scientific™
SH31083.03 |
12.7mm (1/2 in) ID, 3.2mm (1/8 in) wall, C-Flex 374, 5m (16.4 ft) |
Each for $138.00
|
|
|||||
SH3107902
![]() |
Thermo Scientific™
SH31079.02 |
3.2mm (1/8 in) ID, 1.6mm (1/16 in) wall, C-Flex 374, 2.5m (8.2 ft) |
Each for $81.90
|
|
|||||
SH3107903
![]() |
Thermo Scientific™
SH31079.03 |
3.2mm (1/8 in) ID, 1.6mm (1/16 in) wall, C-Flex 374, 5m (16.4 ft) |
Each for $61.00
|
|
|||||
SH3108001
![]() |
Thermo Scientific™
SH31080.01 |
6.35mm (1/4 in) ID, 1.6mm (1/16 in) wall, C-Flex 374, 1m (3.25 ft) |
Each for $33.25
|
|
|||||
SH3108002
![]() |
Thermo Scientific™
SH31080.02 |
6.35mm (1/4 in) ID, 1.6mm (1/16 in) wall, C-Flex 374, 2.5m (8.2 ft) |
Each for $24.75
|
|
|||||
SH3108003
![]() |
Thermo Scientific™
SH31080.03 |
6.35mm (1/4 in) ID, 1.6mm (1/16 in) wall, C-Flex 374, 5m (16.4 ft) |
Each for $97.00
|
|
|||||
SH3108101
![]() |
Thermo Scientific™
SH31081.01 |
6.35mm (1/4 in) ID, 2.4mm (3/32 in) wall, C-Flex 374, 1m (3.25 ft) |
Each for $67.00
|
|
|||||
SH3108102
![]() |
Thermo Scientific™
SH31081.02 |
6.35mm (1/4 in) ID, 2.4mm (3/32 in) wall, C-Flex 374, 2.5m (8.2 ft) |
Each for $77.00
|
|
|||||
SH3108103
![]() |
Thermo Scientific™
SH31081.03 |
6.35mm (1/4 in) ID, 2.4mm (3/32 in) wall, C-Flex 374, 5m (16.4 ft) |
Each for $92.00
|
|
|||||
SH3108201
![]() |
Thermo Scientific™
SH31082.01 |
9.52mm (3/8 in) ID, 3.2mm (1/8 in) wall, C-Flex 374, 1m (3.25 ft)) |
Each for $70.25
|
|
|||||
SH3108202
![]() |
Thermo Scientific™
SH31082.02 |
9.52mm (3/8 in) ID, 3.2mm (1/8 in) wall, C-Flex 374, 2.5m (8.2 ft) |
Each for $90.00
|
|
|||||
SH3107901
![]() |
Thermo Scientific™
SH31079.01 |
3.2mm (1/8 in) ID, 1.6mm (1/16 in) wall, C-Flex 374, 1m (3.25 ft) |
Each for $132.00
|
|
|||||
SH3108203
![]() |
Thermo Scientific™
SH31082.03 |
9.52mm (3/8 in) ID, 3.2mm (1/8 in) wall, C-Flex 374, 5m (16.4 ft) |
Each for $92.40
|
|
|||||
SH3108301
![]() |
Thermo Scientific™
SH31083.01 |
12.7mm (1/2 in) ID, 3.2mm (1/8 in) wall, C-Flex 374, 1m (3.25 ft) |
Each for $42.25
|
|
|||||
SH3108302
![]() |
Thermo Scientific™
SH31083.02 |
12.7mm (1/2 in) ID, 3.2mm (1/8 in) wall, C-Flex 374, 2.5m (8.2 ft) |
Each for $43.25
|
|
|||||
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Approved for storage at ambient conditions (15°C to 30°C). |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.