Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Standard Format HE33 Mini Submarine Unit and Accessories

Catalog No. 45001851 Shop All Cytiva Products
Change view
Click to view available options
HE 33 Mini Submarine Unit; Type: complete
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
45-001-851 HE 33 Mini Submarine Unit; Type: complete
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-851 Supplier Cytiva Supplier No. 80605245

missing translation for 'validMainframeMsg'

Includes

Basic HE 33 Mini Submarine Unit:
Gel running tray, gel casting tray and bubble level

Complete HE 33 Mini Submarine Unit:
Basic unit, 1.5mm thick 8-well comb and comb back

Enhanced passive cooling system allows running gels at high voltages for speed and resolution

  • Coolant-insulated buffer chamber runs 7 x 10cm agarose gels in <20 minutes
  • Convenient, leak-proof casting trays provide fast gel casting without tape
  • UV-transparent gel tray for convenient gel handling and viewing
  • Combs allow up to 32 samples per run
  • Molded base design to be filled with common coolant such as 50% ethylene glycol

Order Info

For Basic unit, combs and comb back should be ordered separately

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Type Mini Submarine Unit (Complete)
Includes Basic unit, 1.5mm thick 8-well comb and comb back
Sample Size Up to 32
Temperature (English) Operating 122°F
Temperature (Metric) Operating 50°C
Volume (English) Buffer 8.4 oz.
Volume (Metric) Buffer 250 mL
Dimensions (L x W x H) 5.2 x 9.5 x 2.8 in. (13 x 24 x 7 cm)
Length (English) 5.2 in.
Length (English) Gel 3.9 in.
Length (Metric) 13 cm
Length (Metric) Gel 10 cm
Width (English) 9.5 in.
Width (English) Gel 2.7 in.
Width (Metric) 24 cm
Width (Metric) Gel 7 cm
Height (English) 2.8 in.
Height (Metric) 7 cm
Voltage 500 V
Wattage 15 w
Amperage 500 mA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.