Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Biotech Supply Standard Hazardous Location (Explosion Proof) Freezer with Natural Refrigerants
GREENER_CHOICESDP

Catalog No. 22651386 Shop All Nor-Lake Scientific Products
Change view
Click to view available options
Capacity (English):
14 cu. ft.
17 cu. ft.
20 cu. ft.
Amperage:
3.5 A
6 A
Min. Clearance:
70 in.
75 in.
80 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Amperage Min. Clearance
22-651-386 14 cu. ft. 3.5 A 75 in.
22-651-387 17 cu. ft. 6 A 70 in.
22-651-388 20 cu. ft. 6 A 80 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-651-386 Supplier American Biotech Supply Supplier No. ABTHCEFP14
Add to Cart
Add to Cart
Request a Quote

Freezers are engineered for cold-storage use in environments where volatile conditions could potentially exist. These units meet NFPA and OSHA guidelines Class 1, Division II, Group C & D for storing substances in hazardous locations.

Standard Hazardous Location (Explosion Proof) Freezers with Natural Refrigerants are engineered for cold-storage use in environments where volatile conditions could potentially exist. All units include a two year parts and labor warranty. They run on natural, hydrocarbon refrigerant for environmental health and economic efficiency. These units meet NFPA and OSHA guidelines Class 1, Division II, Group C & D for storing substances in hazardous locations.*

*Explosion Proof Units are designed for environments where external hazards could exists. For storage of volatile substances in normal laboratory conditions, see ABS's Flammable Storage Units.

  • Microprocessor Temperature Controller
  • Interior shelving
  • Keyed door lock
  • High density urethane foam cabinet and door insulation
  • Not equipped with line cord and plug; must be hardwired in conduit to 120 VAC power source
  • ETL Listed

Specifications

Type HazLoc Freezer
Capacity (English) 14 cu. ft.
Capacity (Metric) 396 L
Temperature Range -15°C to -25°C
Temperature Sensor Microprocessor Temperature Controller
Temperature Stability ±0.1°C
Door Style Solid Swing
No. of Doors 1
Hinge Side Right
Cabinet Material Scratch-resistant Painted Steel
Insulation Polyurethane Foam
Interior Plastic
Exterior Finish Powder-coated Steel
Shelves 6 Fixed
Certifications/Compliance cULus
Style Upright
Display Manual
Color White
Condenser Type Fan-forced
For Use With (Application) Hazardous Location Cold Storage
Ports None
Warranty Two Year Parts and Labor Warranty
Defrost Manual
Min. Clearance 75 in.
Monitoring Options Microprocessor Temperature Controller
Amperage 3.5 A
Plug Type No Plug, Hardwired
Voltage 115 V
Energy Usage 1 to 3 HP
Compressor Bottom-mounted
Refrigerant Natural, Hydrocarbon
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.