Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Standard Solution 120, Reagents

Catalog No. 01336182
Click to view available options
Quantity:
1 L
4 L

Standard Solution 120, Reagent, use for Paints and coatings. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Standard Solution 120
Packaging Poly Bottle
CAS 7732-18-5, 7664-93-9
Specific Gravity 1.0024
Physical Form Liquid
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Fluoride Meter
Solubility Completely miscible
Type Conductivity Standard
Quantity 1 L
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.