Learn More
Bal Supply Standard Taper Joint Clips, Keck Type, POM
Shop All Tip Industries Inc. Products
Click to view available options
Size:
10
12
14
19
24
29
34
40
45
Color:
Blue
Brown
Gold
Green
Orange
Red
Violet
Yellow
Description
- Made from Polyoxymethylene (POM)
- Will not scratch glass
- Color-coded
- For standard taper joints
Specifications
Specifications
Autoclavable | Autoclavable |
Quantity | 10 Pk. |
Type | Standard Taper Joint Clip |
Material | POM |
Size | 29 |
Color | Red |
Jaw Type | Standard Taper |
Sterility | Non-sterile |