Description
- 100% virgin spunbonded, non-linting polypropylene
- Trimmed in elastic for a snug, adjustable fit
- Elastic seam is turned in to reduce particle generation
- With a non-marking, latex-based, anti-skid bottom surface
- Color: sky blue
Order Info
100 Each per vacuum-packed, heat-sealed polybag; three bags per case.
Compliance
Conform to test standard IES-RP-CC-003.2
Specifications
Specifications
| Size | Large |
| Material | Polypropylene |
| Color | Sky Blue |
| Closure Type | Elastic Ankle |
| Product Type | Controlled Environments Shoe Covers |