Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stanniocalcin 1/STC-1 Antibody (SR1521), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32224750UL
Description
Stanniocalcin 1/STC-1 Monoclonal antibody specifically detects Stanniocalcin 1/STC-1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Stanniocalcin 1/STC-1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
SR1521 | |
Western Blot 1:500-1:200, Immunoprecipitation 1:50 | |
stanniocalcin 1, stanniocalcin-1, STCSTC-1 | |
A synthesized peptide derived from human Stanniocalcin 1/STC-1 (Uniprot #: P52823) | |
50 μg | |
Signal Transduction | |
6781 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction