Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanniocalcin 2 Mouse anti-Human, Mouse, Clone: 4H1E7, Proteintech
SDP

Catalog No. 501736217 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6217 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6217 Supplier Proteintech Group Inc Supplier No. 600631IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Has an anti-hypocalcemic action on calcium and phosphate homeostasis.

Specifications

Antigen Stanniocalcin 2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4H1E7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene STC2
Gene Accession No. O76061, O88452
Gene Alias stanniocalcin 2, Stanniocalcin related protein, STC 2, STC related protein, STC2, STCRP
Gene Symbols STC2
Host Species Mouse
Immunogen Stanniocalcin 2 Fusion Protein Ag0359
Purification Method Caprylic Acid Precipitation
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20856, 8614
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.