Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stannous Chloride, 40% (w/v) in concentrated Hydrochloric Acid, Ricca Chemical

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
Specifications
Specifications
Color | Colorless to Green-Yellow |
CAS Min % | 73.0, 24.82 |
CAS Max % | 76.0, 25.84 |
Percent Purity | 40% |
Grade | Laboratory |
Packaging | Amber Glass Bottle |
Solvent or Matrix | Concentrated Hydrochloric Acid |
pH | <1 |
Quantity | 500 mL |
Concentration or Composition Notes | 40% (w/v) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction