Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride Anhydrous (Crystalline Powder or Flakes/Technical), Fisher Chemical™

Catalog No. p-34522 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
Synonym Tin (II) Chloride Anhydrous
PubChem CID 24479
ChEBI CHEBI:78067
IUPAC Name λ²-tin(2+) dichloride
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Boiling Point 652°C
Melting Point 246°C
Color White
pH 2
Physical Form Solid
Quantity 100 g
Formula Weight 189.62
Grade Technical
Identification Pass Test
Packaging Glass Bottle
Density 3.950g/cm³
Chemical Name or Material Stannous Chloride Anhydrous
Show More Show Less
DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.