Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride AR Crystal, Macron Fine Chemicals™

Catalog No. p-3178343
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10025-69-1
Molecular Formula SnCl2·2H2O

Specifications

Physical Form Crystalline
Quantity 500 g
Formula Weight 225.63
Packaging POLYSTORMOR
Chemical Name or Material Stannous Chloride, Dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.