Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Stannous Chloride Dihydrate
SCI_ED

Catalog No. S25578 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Lab Grade
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25578 25 g Lab Grade
S25578B 500 g Lab Grade
S25578A 100 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25578
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10025-69-1
Synonym Tin (II) Chloride Dihydrate

Specifications

Color Colorless
Quantity 25 g
Assay Percent Range >95%
Linear Formula SnCl2·2H2O
Formula Weight 225.63
Grade Laboratory
Packaging Lab Grade
Chemical Name or Material Stannous Chloride, Dihydrate
Safety handling Check Alert storage code Red
ShelfLife 1 year
DOTInformation DOT Class 8, III, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.