Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride, Dihydrate, Crystal, Reagent, ACS, 98-103%, Spectrum™ Chemical
SDP

Catalog No. 18606992
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

S1535, 10025-69-1, SnCl2·2H2O

Spectrum™ Chemical Stannous Chloride, Dihydrate, Crystal, Reagent, ACS is also known as Tin Chloride, has a white solid crystal like aspect. Mostly used in electrolytic baths destined for tin-plating, it is also widely used as a reducing agent. In the textile industry it is employed as a mordant to increase the brightness of certain colors in the fabric. It is also employed by metal enthusiasts to determine platinum and gold group metals. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2H4O2Sn
Molecular Weight (g/mol) 225.64
InChI Key FWPIDFUJEMBDLS-UHFFFAOYSA-L
IUPAC Name λ²-tin(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Sn++]

Specifications

CAS Max % 1
Quantity 12 kg
Percent Purity 98 to 103%
Grade ACS
Packaging Poly Pail
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.