Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride Reagent I, Ricca Chemical
SDP

Catalog No. 79974 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7997-4 120 mL Natural Poly Bottle
7997-16 500 mL Natural Poly Bottle
R79970001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 7997-4 Supplier Ricca Chemical Company Supplier No. 79974
Add to Cart
Add to Cart

For Phosphorus (Phosphate) Analysis (without extraction)

Chemical Identifiers

CAS 56-81-5 , 10025-69-1
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Color Colorless
CAS Min % 96.0, 1.9
CAS Max % 100.0, 1.98
Grade Laboratory
Packaging Natural Poly Bottle
pH Neutral
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Stannous Chloride Reagent I
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.