Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride Suspension, 10% (w/v) in 0.5 N Sulfuric Acid, Ricca Chemical
SDP

Catalog No. 7996532 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79965-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 79965-32 Supplier Ricca Chemical Company Supplier No. 7996.532
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 10025-69-1 , 7664-93-9
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

CAS Min % 87.0, 8.71, 2.24
CAS Max % 91.0, 9.07, 2.33
Physical Form Liquid
Quantity 1 L
Percent Purity 10% W/V
Grade Laboratory
Packaging Amber Glass Bottle
Chemical Name or Material Stannous Chloride, 10% w/v in 0.5Normal Sulfuric Acid Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.