Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAP1 Polyclonal Antibody, Abbomax

Catalog No. 502204622
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4622 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4622 Supplier Abbomax Inc Supplier No. 630820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Tonsil Cellular location: Nucleus and cytoplasm.

STAP1 Polyclonal antibody specifically detects STAP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen STAP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene STAP1
Gene Accession No. Q9JM90, Q9ULZ2
Gene Alias AI586015; BCR downstream signaling 1; BCR downstream-signaling protein 1; BRDG1; docking protein BRDG1; signal transducing adaptor family member 1; signal-transducing adaptor protein 1; Stap1; STAP-1; Stem cell adaptor protein 1; stem cell adaptor protein STAP-1
Host Species Rabbit
Immunogen A synthetic peptide LHE VLE REK KRR IET derived from human STAP1
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26228, 305269, 56792
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.