Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAP2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NB300987
Description
STAP2 Polyclonal specifically detects STAP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
STAP2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
BKSbrk kinase substrate, Breast tumor kinase substrate, BRK substrate, FLJ20234, signal transducing adaptor family member 2, signal-transducing adaptor protein 2, signal-transducing adaptor protein-2, STAP-2 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize isoforms 1 (NP_060190.2) and isoform 2 (NP_001013863.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.05 - 0.3 ug/ml, Immunohistochemistry 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, Peptide ELISA Detection limit 1:16000 | |
Q9UGK3 | |
STAP2 | |
Peptide with sequence C-ELQKKLEKRRALEH corresponding to C-Terminus according to NP_060190.2, NP_001013863.1. | |
0.1 mg | |
Signal Transduction | |
55620 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction