Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STAP2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537984
Description
This antibody is predicted to react with rat and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene encodes the substrate of breast tumor kinase, an Src-type non-receptor tyrosine kinase. The encoded protein possesses domains and several tyrosine phosphorylation sites characteristic of adaptor proteins that mediate the interactions linking proteins involved in signal transduction pathways. Alternative splicing results in multiple transcript variants.
Specifications
STAP2 | |
Polyclonal | |
Unconjugated | |
STAP2 | |
AW049765; BKS; Breast tumor kinase substrate; brk kinase substrate; BRK substrate; FLJ20234; signal transducing adaptor family member 2; signal-transducing adaptor protein 2; signal-transducing adaptor protein-2; STAP2; STAP-2 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
106766 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8R0L1 | |
STAP2 | |
Peptide with sequence C-EEKLQKRRALEH, from the C-terminus of mouse Stap2 (aa 400-411). | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction