Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575649
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575649 Supplier Thermo Scientific Supplier No. PA575649
Add to Cart
Add to Cart

STAP2 Polyclonal Antibody

STAP2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen STAP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene STAP2
Gene Accession No. Q8R0L1, Q9UGK3
Gene Alias AW049765; BKS; Breast tumor kinase substrate; brk kinase substrate; BRK substrate; FLJ20234; signal transducing adaptor family member 2; signal-transducing adaptor protein 2; signal-transducing adaptor protein-2; STAP2; STAP-2
Gene Symbols STAP2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human STAP2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106766, 363334, 55620
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.