Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582014 Supplier Invitrogen™ Supplier No. PA582014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NYFVSHTKKA LVPFLLDEDY EKVLGYVEAD KENGENVWVA PSAPGPGPAP CTGGPKPLSP ASSQDKLPPL PPLPNQEENY VTPIGDGPAV DYENQDVASS S Highest antigen sequence indentity to the following orthologs: Rat - 60%, Mouse - 61%.

This gene encodes the substrate of breast tumor kinase, an Src-type non-receptor tyrosine kinase. The encoded protein possesses domains and several tyrosine phosphorylation sites characteristic of adaptor proteins that mediate the interactions linking proteins involved in signal transduction pathways. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STAP2
Gene Accession No. Q9UGK3
Gene Alias AW049765; BKS; Breast tumor kinase substrate; brk kinase substrate; BRK substrate; FLJ20234; signal transducing adaptor family member 2; signal-transducing adaptor protein 2; signal-transducing adaptor protein-2; STAP2; STAP-2
Gene Symbols STAP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STAP2. Recombinant protein control fragment (Product #RP-102140).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55620
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.