Learn More
3M™ Petrifilm™ Staph Express Disk, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. p-7232132
Click to view available options
Quantity:
100 Pack
20 Pack
Used to identify Staphylococcus aureus from all suspect colonies for samples with high background flora
  • Utilizes convenience and efficiency of the 3M™ Petrifilm™ Plate platform
  • Proven cost effective tool for detection of Staph. aureus
  • Results in as few as 22 hours
Specifications
Detects Staphylococcus aureus
Product Type Disk
Test Type Animal Feed, Pet Food
For Use With (Application) Identifying Staphylococcus aureus from suspect colonies with high background flora
Quantity 20 Pack
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.