Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Staphylococcus aureus Enolase Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117601
Description
This antibody has been tested in direct-ELISA.
Catalyzes the reversible conversion of 2-phosphoglycerate into phosphoenolpyruvate. It is essential for the degradation of carbohydrates via glycolysis.
Specifications
Staphylococcus aureus Enolase | |
Polyclonal | |
Unconjugated | |
2-phospho-D-glycerate hydro-lyase; 2-phosphoglycerate dehydratase; eno; Enolase | |
Recombinant Staphylococcus aureus Enolase protein (1-434aa). | |
100 μg | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction