Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Staphylococcus Aureus Enterotoxin B Antibody (S222), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1108272
Description
Staphylococcus Aureus Enterotoxin B Monoclonal specifically detects Staphylococcus Aureus Enterotoxin B in Bacteria samples. It is validated for Immunoassay, Sandwich ELISA.Specifications
Staphylococcus Aureus Enterotoxin B | |
Monoclonal | |
Unconjugated | |
S. aureus | |
Recombinant St. aureus enterotoxin B expressed in E.coli | |
0.2 mg | |
Virology Bacteria and Parasites | |
Clone S222 reacts with enterotoxin B of St. aureus but does not cross-react with St.aureus enterotoxins A, C1, C2, C3, D and E. | |
Store at 4C. Do not freeze. | |
IgG1 |
Immunoassay, ELISA | |
S222 | |
Immunoassay, Sandwich ELISA 1:100-1:2000 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction