Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Staphylococcus Aureus Enterotoxin B Antibody (S643), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073025UV
Description
Staphylococcus Aureus Enterotoxin B Monoclonal specifically detects Staphylococcus Aureus Enterotoxin B in Bacteria samples. It is validated for ELISA, Sandwich ELISA.Specifications
Staphylococcus Aureus Enterotoxin B | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
RUO | |
Staphylococcus Aureus Enterotoxin B. This does not cross react with S. aureus enterotoxins A, C, D and E. | |
Store at 4C in the dark. | |
IgG1 |
ELISA | |
S643 | |
ELISA, Sandwich ELISA | |
S. aureus | |
Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant S. aureus enterotoxin B expressed in E. coli. | |
0.2 mL | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction