Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Staphylococcus aureus (Methicillin-resistant) Monoclonal Antibody (332/423)
GREENER_CHOICE

Catalog No. MA183467
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA183467 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA183467 Supplier Invitrogen™ Supplier No. MA183467
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

For IHC applications, this antibody reacts with isolates fixed by phenol, formalin and boiling. Mouse anti methicillin resistant Staphylococcus aureus antibody, clone 332/43 recognizes methicillin-resistant Staphylococcus aureus (MRSA), also known as oxacillin-resistant Staphylococcus aureus (ORSA).

Staphylococcus aureus is a type of bacteria that commonly lives on the skin and in the nasal passages of healthy individuals. However, it can also cause a range of infections, from minor skin infections to more serious conditions such as pneumonia, sepsis, and endocarditis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Staphylococcus aureus (Methicillin-resistant)
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 332/423
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias MRSA; s aureus; S. aureus; staphylococcus aureus methicillin-resistant
Host Species Mouse
Immunogen Native bacterium.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.