Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Staphylococcus aureus Monoclonal Antibody (Staph12-569.3), Invitrogen™

Catalog No. PIMA546041
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546041 100 μg
01-671-941 500 μg
PIMA546117 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546041 Supplier Invitrogen™ Supplier No. MA546041
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10710 detects Staphylococcus aureus from Staph. aureus and Staph. epidermidis samples. MA1-10710 has been successfully used in ELISA applications. The MA1-10710 immunogen is UV-inactivated Staphylococcus aureus cells.

Staphylococcus aureus is a type of bacteria that commonly lives on the skin and in the nasal passages of healthy individuals. However, it can also cause a range of infections, from minor skin infections to more serious conditions such as pneumonia, sepsis, and endocarditis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Staphylococcus aureus
Applications ELISA
Classification Monoclonal
Clone Staph12-569.3
Concentration 2.8 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias MRSA; s aureus; S. aureus; staphylococcus aureus methicillin-resistant
Host Species Mouse
Immunogen UV-inactivated Staphylococcus aureus cells.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.