Learn More
Invitrogen™ Staphylococcus aureus Polyclonal Antibody, Biotin

Description
The PA1-73174 antibody reacts with Staphylococcus aureus. PA1-73174 has been successfully used in Immunofluorescence.Immunohistochemistry, ELISA, and Western Blot applications are recommended but have not been tested. The PA1-73174 antibody was raised against soluble and structural antigens of the whole bacterium; ATCC #27660. Short term storage, up to 6 months, is liquid at 4ºC. For longer term storage, aliquot and freeze.
Specifications
Specifications
| Antigen | Staphylococcus aureus |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | MRSA; s aureus; S. aureus; staphylococcus aureus methicillin-resistant |
| Host Species | Rabbit |
| Immunogen | Staphylococcus aureus whole cells. |
| Purification Method | Protein A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.