Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Staphylococcus aureus PBP2a (Methicillin-resistant) Monoclonal Antibody (AC10), Biotin
GREENER_CHOICE

Catalog No. PIMA546569
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546569 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546569 Supplier Invitrogen™ Supplier No. MA546569
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Staphylococcus aureus PBP2a (Methicillin-resistant) Monoclonal Antibody for ELISA, ICC/IF

Methicillin-resistant Staphylococcus aureus (MRSA) is a type of bacteria that is resistant to the antibiotic methicillin. The resistance is due to the presence of a protein called penicillin-binding protein 2a (PBP2a), which is encoded by the mecA gene. PBP2a has a low affinity for methicillin and other beta-lactam antibiotics, which allows the bacteria to survive and multiply in the presence of these drugs. A target for PBP2a would be a transpeptidase enzyme that is responsible for the synthesis of the bacterial cell wall and is a key factor in the resistance of MRSA to beta-lactam antibiotics.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Staphylococcus aureus PBP2a (Methicillin-resistant)
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone AC10
Concentration 1 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide
Gene Alias MRSA
Host Species Mouse
Immunogen Native Staphylococcus aureus (methicillin resistant).
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.