Learn More
Invitrogen™ Staphylococcus Enterotoxin A Monoclonal Antibody (D99G)

Description
The MA1-7480 antibody reacts with Staphylococcus Enterotoxin toxin A capture in bacterial and infected tissue samples. See related products for paired antibody. MA1-7480 has been successfully used in ELISA applications. The MA1-7480 antibody was raised against Staphylococcus Toxin A.
Specifications
Specifications
| Antigen | Staphylococcus Enterotoxin A |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | D99G |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Host Species | Mouse |
| Immunogen | Staph. Toxin A. |
| Purification Method | Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.