Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staphylococcus Enterotoxin A Monoclonal Antibody (J98J), Invitrogen™

Catalog No. PIMA17481
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17481 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17481 Supplier Thermo Scientific Supplier No. MA17481

Mouse Monoclonal Antibody

The MA1-7481 antibody reacts with Staphylococcus Enterotoxin toxin A probe in bacterial and infected tissue samples. See related products for paired antibody. MA1-7481 has been successfully used in ELISA applications. The MA1-7481 antibody was raised against Staphylococcus Toxin A.

MA1-74347 recognises Staphylococcus aureus enterotoxin type A. Staphylococcus aureus produces several different types of enterotoxins (SE’s). The SE’s are short, soluble proteins that are highly stable and resistant to most proteolytic enzymes. They belong to a family of pyrogenic toxins and as such cause an increase in body temperature, immunosuppression and non-specific T-cell proliferation. These activities are referred to as superantigen activity and results from the interaction of SE’s with T-cell antigen receptors and the major histocompatibility complex (MHC) on antigen presenting cells. In addition to their superantigen activity, SE’s are also emetics (they cause vomiting). The SE’s are important in food poisoning. Enterotoxin A is one of the SE’s most commonly identified in clinical isolates. MA1-74347 may be used as a capture antibody in ELISA assays for S. aureus enterotoxin A in combination with MCA2848 as a detection agent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Staphylococcus Enterotoxin A
Applications ELISA
Classification Monoclonal
Clone J98J
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Host Species Mouse
Immunogen Staph. Toxin A.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.