Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Staphylococcus Enterotoxin B Monoclonal Antibody (H80J)
GREENER_CHOICE

Catalog No. MA17479
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17479 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17479 Supplier Invitrogen™ Supplier No. MA17479
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7479 antibody reacts with Staphylococcus Enterotoxin toxin B probe in bacterial and infected tissue samples. See related products for paired antibody. MA1-7479 has been successfully used in ELISA applications. The MA1-7479 antibody was raised against Staphylococcus Toxin B.

Staphylococcus aureus Enterotoxin B consists of 239 amino acids with a molecular weight of ~28 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Staphylococcus Enterotoxin B
Applications ELISA
Classification Monoclonal
Clone H80J
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Host Species Mouse
Immunogen Staph. Toxin B.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.