Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staphylococcus Enterotoxin pan Monoclonal Antibody (S13), Invitrogen™

Catalog No. PIMA183468
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA183468 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA183468 Supplier Thermo Scientific Supplier No. MA183468

Mouse Monoclonal Antibody

MA1-83468 is specific for the Staphylococcus enterotoxins, and has shown the following reactivities when tested in ELISA: enterotoxin B 100%, enterotoxin C1 18%, enterotoxin C2 82%, enterotoxin D 49%, enterotoxin E 24%. Mouse anti Staphylococcus Enterotoxin A/B/C1/C2/D/E antibody, clone S13 Staphylococcus enterotoxin. It has shown the following reactivities when tested in ELISA. enterotoxin A65%enterotoxin B100%enterotoxin C118%enterotoxin C282%enterotoxin D49%enterotoxin E24%.

Staphylococcus Enterotoxin pan Monoclonal antibody specifically detects Staphylococcus Enterotoxin pan in Bacteria samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Staphylococcus Enterotoxin pan
Applications ELISA
Classification Monoclonal
Clone S13
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide
Host Species Mouse
Immunogen Purified enterotoxin A from FRI-722H strain of S.aureus.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.