Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Staphylococcus epidermidis Monoclonal Antibody (17-5)
GREENER_CHOICE

Catalog No. 01672092
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-092 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-092 Supplier Invitrogen™ Supplier No. MA135788
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody reacts with several Staphylococcus strains using ELISA. It does not react with several members of the Enterobacteriaceae and not with S. Aureus. It is not tested for IHC-P. The epitope is not determined, but it might be a component of the cell wall. The antibody recognizes whole bacteria.

Although pathogenic bacteria constitute only a small portion of bacterial species, infectious diseases caused by pathogenic bacteria affect millions of people worldwide. Diarrheal diseases, many of which are bacterial, are the second leading cause of death worldwide, claiming an estimated 5 million people annually. Furthermore, an estimated 3 million people die of tuberculosis annually, 500,000 people die of pertussis, and 25,000 die of typhoid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Staphylococcus epidermidis
Applications ELISA
Classification Monoclonal
Clone 17-5
Concentration 200 μg/mL
Conjugate Unconjugated
Formulation tissue culture supernatant, cell culture media with 0.02% sodium azide
Gene Alias s epidermidis; S. epidermidis
Host Species Mouse
Immunogen Proprietary Immunogen.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.