Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595765
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595765 Supplier Invitrogen™ Supplier No. PA595765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: K-562, Mouse ovary, Rat testis; Cellular Location: Mitochondrion Immunogen sequence: EETLYSDQEL AYLQQGEEAM QKALGILSNQ EGWKKESQQD NGDKVMSKVV PDVGKVFRLE VVVDQPMERL YEELVERMEA MGEWNPNVKE IKVLQKIGKD TFITHELAAE AAGNLVGPRD FVSVRCAKRR GSTCVLAGMA TDFGNMPEQK GVIRAEHGPT CMVLHPLAGS PSKTKLTWLL SIDLKGWLPK SIINQVLSQT QVDFANHLRK RLESHPASEA RC.

Steroidogenic acute regulatory protein (StAR) plays a significant role in cholesterol transport from the cytoplasmic outer membrane to the inner mitochondrial membrane. The 37 kDa precursor is cleaved to generate an active 28 kDa protein capable of facilitating cholesterol metabolism into pregnenolone. StAR is prevalently expressed in mitochondria of steroid-producing adrenal and gonadal tissue. Abnormalities in StAR gene expression are impacted in autosomal Lipoid Congenial Adrenal Hyperplasia (LCAH) resulting in defects in pregnenolone and cortisol synthesis. The mechanism of cholesterol binding to StAR has yet to be elucidated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAR
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene STAR
Gene Accession No. P49675, P51557, P97826
Gene Alias AV363654; cholesterol trafficker; D8Ertd419e; lipid transporter; luteinizing hormone-induced protein; mitochondrial steroid acute regulatory protein; STAR; StAR related lipid transfer (START) domain containing 1; stARD1; StAR-related lipid transfer (START) domain containing 1; START domain containing 1; START domain-containing protein 1; steroid acute regulatory protein; steroidogenic acute regulator; steroidogenic acute regulatory protein; steroidogenic acute regulatory protein, mitochondrial; testis secretory sperm-binding protein Li 241mP
Gene Symbols STAR
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20845, 25557, 6770
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.