Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Starch
SCI_ED

Catalog No. s25582 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Powder - Soluble Starch - Lab Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 9005-25-8
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00082026,MFCD00132834
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC Name 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Color White
Assay Percent Range 0.1% w/v
Linear Formula (C6H10O5)n
Density 1.5g/mL
Quantity 100 g
Packaging Powder - Soluble Starch - Lab Grade
Chemical Name or Material Starch
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.