Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch, Soluble, Powder, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18607001 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

S1555, 9005-84-9, (C6H10O5)n

Spectrum™ Chemical Starch, Soluble, Powder, Reagent, ACS is indicator for redox titrations where triiodide is present. Starch forms a very dark blue-black complex with triiodide which can be made by mixing iodine with iodide (often from potassium iodide). The color of the starch complex is so deep, that it can be detected visually when the concentration of the iodine is as low as 0.00002 M at 20 °C. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 9005-84-9
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
InChI Key GUBGYTABKSRVRQ-ASMJPISFSA-N
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

CAS Max % 1
pH 5.0 to 7.0
Ignition Residue 0.004
Packaging Poly Bottle
Quantity 125 g
Grade ACS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.