Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Starch Indicator, 0.3% w/v, Reagents
Click to view available options
Quantity:
1 L
Specifications
Specifications
Chemical Name or Material | Starch Indicator, 0.3% w/v |
CAS | 7732-18-5, 9005-84-9, 69-72-7 |
Boiling Point | 100°C |
Percent Purity | 0.3% |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.002 |
Color | White |
pH | 5 to 7 |
Quantity | 1 L |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction