Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch, Soluble, ACS Reagent, Reagents

Catalog No. 01337653
Click to view available options
Quantity:
125 g
500 g
2.5 kg

Starch, Soluble, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 9005-84-9
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
InChI Key GUBGYTABKSRVRQ-ASMJPISFSA-N
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Boiling Range 668°C
Color White
Boiling Point 668°C
Packaging Poly Bottle
Quantity 2.5 kg
Specific Gravity 1.5
Grade ACS
Physical Form Solid
Chemical Name or Material Starch, Soluble, ACS Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.