Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Starch Solution, 1% w/v Preserved with Chloroform, Reagents
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Cubitainer
Poly Bottle
Specifications
Specifications
| Chemical Name or Material | Starch Solution, 1% w/v Preserved with Chloroform |
| Boiling Point | 100°C |
| CAS | 7732-18-5, 9005-84-9, 67-66-3 |
| Percent Purity | 1% |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.005 |
| Color | White |
| Quantity | 500 mL |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction