Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Starch Solution, 2% w/v, Mercury-Free, with Salicylic Acid Preservative, Reagents
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Specifications
Specifications
Chemical Name or Material | Starch Solution, 2% w/v, Mercury-Free, with Salicylic Acid Preservative |
CAS | 7732-18-5, 9005-84-9, 69-72-7 |
Boiling Point | 100°C |
Percent Purity | 2% |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.02 |
Color | White |
Quantity | 1 L |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction