Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STARD10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550174
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550174 Supplier Invitrogen™ Supplier No. PA550174
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total STARD10 protein.

STARD10 gene ontology annotations related to this gene include bile acid secretion; positive regulation of peroxisome proliferator activated receptor signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STARD10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene STARD10
Gene Accession No. Q9JMD3, Q9Y365
Gene Alias Antigen NY-CO-28; AV048538; CGI52; CGI-52; NY-C0-28; NY-CO-28; PCTP2; PC-TP2; Pctpl; PCTP-L; PCTP-like protein; Sdccag28; Sdccagg28; serologically defined colon cancer antigen 10; serologically defined colon cancer antigen 28; Serologically defined colon cancer antigen 28 homolog; StAR related lipid transfer domain containing 10; STARD10; StAR-related lipid transfer (START) domain containing 10; StAR-related lipid transfer domain containing 10; stAR-related lipid transfer protein 10; START domain containing 10; START domain-containing protein 10; TISP-81
Gene Symbols STARD10
Host Species Rabbit
Immunogen Synthesized peptide derived from N-terminal of human STARD10.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10809, 293150, 56018
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.