Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STARD13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558831
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558831 Supplier Thermo Scientific Supplier No. PA558831
Add to Cart
Add to Cart

STARD13 Polyclonal Antibody

STARD13 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen STARD13
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STARD13
Gene Accession No. Q9Y3M8
Gene Alias 46H23.2; ARHGAP37; deleted in liver cancer 2 protein; DLC2; DLC-2; GT650; LINC00464; long intergenic non-protein coding RNA 464; Rho GTPase activating protein on chromosome 13q12; Rho GTPase-activating protein; serologically defined colon cancer antigen 13; serologically defined colon cancer antigen 28; STA13; StAR related lipid transfer domain containing 13; STARD13; StAR-related lipid transfer (START) domain containing 13; StAR-related lipid transfer domain containing 13; stAR-related lipid transfer protein 13; START domain-containing protein 13
Gene Symbols STARD13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STARD13. Recombinant protein control fragment (Product #RP-97949).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90627
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.