Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STARD13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563622
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563622 Supplier Invitrogen™ Supplier No. PA563622
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSDEEDLCIS NKWTFQRTSR RWSRVDDLYT LLPRGDRNGS PGGTGMRNTT SSESVLTDLS EPEVCSIHSE SSGGSDSRSQ PGQCC Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.

This gene encodes a protein which contains an N-terminal sterile alpha motif (SAM) for protein-protein interactions, followed by an ATP/GTP-binding motif, a GTPase-activating protein (GAP) domain, and a C-terminal STAR-related lipid transfer (START) domain. It may be involved in regulation of cytoskeletal reorganization, cell proliferation, and cell motility, and acts as a tumor suppressor in hepatoma cells. The gene is located in a region of chromosome 13 that is associated with loss of heterozygosity in hepatocellular carcinomas. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STARD13
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STARD13
Gene Accession No. Q9Y3M8
Gene Alias 46H23.2; ARHGAP37; deleted in liver cancer 2 protein; DLC2; DLC-2; GT650; LINC00464; long intergenic non-protein coding RNA 464; Rho GTPase activating protein on chromosome 13q12; Rho GTPase-activating protein; serologically defined colon cancer antigen 13; serologically defined colon cancer antigen 28; STA13; StAR related lipid transfer domain containing 13; STARD13; StAR-related lipid transfer (START) domain containing 13; StAR-related lipid transfer domain containing 13; stAR-related lipid transfer protein 13; START domain-containing protein 13
Gene Symbols STARD13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STARD13. Recombinant protein control fragment (Product #RP-104223).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90627
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.