Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STARD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561644
Description
Immunogen sequence: CRLDWDSLMT SLDILENFEE NCCVMRYTTA GQLWNIISPR EFVDFSYTVG YKEGLLSCGI SLDWDEKRPE FVRGY Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.
STARD4 may be involved in the intracellular transport of sterols or other lipids, and may bind cholesterol or other sterols.
Specifications
STARD4 | |
Polyclonal | |
Unconjugated | |
STARD4 | |
4632419C16Rik; 9030213J02Rik; AA517649; AW324468; StAR related lipid transfer domain containing 4; Stard4; StAR-related lipid transfer (START) domain containing 4; StAR-related lipid transfer domain containing 4; stAR-related lipid transfer protein 4; START domain containing 4, sterol regulated; START domain-containing protein 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
134429 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96DR4 | |
STARD4 | |
Recombinant protein corresponding to Human STARD4. Recombinant protein control fragment (Product #RP-96949). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction