Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STARD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548459
Description
STARD5 Polyclonal Antibody for Western Blot, IHC (P), Flow
May be involved in the intracellular transport of sterols or other lipids. May bind cholesterol or other sterols.
Specifications
STARD5 | |
Polyclonal | |
Unconjugated | |
STARD5 | |
18B7-T7(GS); 2310058G22Rik; AI607880; D7Ertd152e; RGD1561783; StAR related lipid transfer domain containing 5; Stard5; StAR-related lipid transfer (START) domain containing 5; StAR-related lipid transfer domain containing 5; stAR-related lipid transfer protein 5; StAR-related protein 1-4E; StAR-related protein p3-15E/p3-16E; START domain containing 5; START domain-containing protein 5 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
80765 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9NSY2 | |
STARD5 | |
KLH conjugated synthetic peptide between 26-55 amino acids from the N-terminal region of human STARD5. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction